Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026147.5 | NP_080423.1 | 2 | 168 | |
GenBank mRNA | AK002405.1 | - | 2 | 169 | |
AK014594.1 | - | 168 | |||
AK136398.1 | - | 2 | 171 | ||
AK145656.1 | - | 2 | 169 | ||
AK150867.1 | - | 2 | 168 | ||
AK150972.1 | - | 2 | 168 | ||
BC011323.1 | - | 2 | 155 | 3257215 | |
BC090389.1 | - | 2 | 159 | 3482252 |