Gene Transcripts

Gene Symbol
MC1R
Entrez Gene ID
4157
Gene Name
melanocortin 1 receptor
Gene Aliases
CMM5, MSH-R, SHEP2
Chromosome Location
Chr. 16: 89917879 - 89920977 on Build GRCh38
UniGene ID
Hs.513829
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002386.3 NP_002377.4 1 1602
GenBank mRNA AK024232.1 - 1 1604
AK054666.1 - 2 572
AK225902.1 - 1 619
AK311630.1 - 2436
BC007856.2 - 1 601 4298428
BC080622.1 - 1 428 6267157
X67594.1 - 1 390

View Full Product Details