Gene Transcripts

Gene Symbol
Slc38a2
Entrez Gene ID
67760
Gene Name
solute carrier family 38, member 2
Gene Aliases
5033402L14Rik, AI316867, ATA2, SAT2, mKIAA1382
Chromosome Location
Chr. 15: 96687386 - 96699724 on Build GRCm38
UniGene ID
Mm.46754
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175121.3 NP_780330.2 14 1648
GenBank mRNA AK031615.1 - 14 1679
AK033812.1 - 2 188
AK129342.1 - 14 1609
AK164568.1 - 8 5411
AK169378.1 - 4 407
BC037675.1 - 11 1055
BC041108.1 - 14 1518 4038546
BC048178.1 - 14 1648 6825801
BC049271.1 - 7 671
BC057454.1 - 1 29

View Full Product Details