Gene Transcripts

Gene Symbol
Bambi
Entrez Gene ID
68010
Gene Name
BMP and activin membrane-bound inhibitor
Gene Aliases
2610003H06Rik
Chromosome Location
Chr. 18: 3507957 - 3516404 on Build GRCm38
UniGene ID
Mm.284863
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026505.2 NP_080781.1 2 419
GenBank mRNA AF153440.1 - 2 405
AK011305.1 - 2 442
AK017303.1 - 2 421
AK045192.1 - 2 419
AK143185.1 - 2 433
AK164632.1 - 2 422
BC019378.1 - 2 401 5009872

View Full Product Details