Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_197985.3 | NP_932102.2 | 3 | 409 | |
GenBank mRNA | AK039667.1 | - | 3 | 617 | |
AK046591.1 | - | 3 | 416 | ||
AK134787.1 | - | 3 | 425 | ||
AK148295.1 | - | 3 | 551 | ||
AK160534.1 | - | 3 | 426 | ||
AK161404.1 | - | 3 | 374 | ||
AY424291.1 | - | 3 | 409 | ||
BC024094.1 | - | 3 | 409 | 5132696 | |
BC064109.1 | - | 3 | 407 | 3371697 |