Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001099675.1 | NP_001093145.1 | 2 | 393 | |
NM_001134646.1 | NP_001128118.1 | 3 | 443 | ||
GenBank mRNA | AA423234.1 | - | 2 | 393 | |
AK003822.1 | - | 2 | 389 | ||
CA466100.1 | - | 3 | 435 |