Gene Transcripts

Gene Symbol
MEOX2
Entrez Gene ID
4223
Gene Name
mesenchyme homeobox 2
Gene Aliases
GAX, MOX2
Chromosome Location
Chr. 7: 15611212 - 15686683 on Build GRCh38
UniGene ID
Hs.170355
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005924.4 NP_005915.2 1 451
GenBank mRNA AK313386.1 - 1 379
BC017021.1 - 1 351 3917118
CR536553.1 - 1 170
DA586635.1 - 1 451
X82629.1 - 1 435

View Full Product Details