Gene Transcripts

Gene Symbol
Ms4a6d
Entrez Gene ID
68774
Gene Name
membrane-spanning 4-domains, subfamily A, member 6D
Gene Aliases
1110058E16Rik, AI447446, Ms4a11
Chromosome Location
Chr. 19: 11586606 - 11604804 on Build GRCm38
UniGene ID
Mm.290390
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026835.2 NP_081111.1 5 547
GenBank mRNA AB026047.1 - 5 480
AF237911.1 - 5 509
AK004295.1 - 5 530
AK152859.1 - 5 527
AK156878.1 - 5 530
AY039037.1 - 5 509
BC018331.1 - 5 484 3964037
BC029738.1 - 5 491 4016611
BG087418.2 - 5 547

View Full Product Details