Gene Transcripts
- Gene Symbol
- Zg16
- Entrez Gene ID
- 69036
- Gene Name
- zymogen granule protein 16
- Gene Aliases
- 1810010M01Rik, AI593689, ZG16p
- Chromosome Location
- Chr. 7: 127050156 - 127052701 on Build GRCm38
- UniGene ID
- Mm.21835
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_026918.2 |
NP_081194.1
|
2, 3 |
185 |
|
GenBank mRNA |
BC031800.1 |
- |
2, 3 |
185 |
|
View Full Product Details