Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026987.2 | NP_081263.2 | 13 | 2196 | |
GenBank mRNA | AK122314.2 | - | 13 | 2131 | |
BC009147.1 | - | 13 | 2155 | 2647184 | |
BC046781.1 | - | 13 | 2124 | 5716182 | |
BC138204.1 | - | 13 | 2135 | 8861223 | |
BC138205.1 | - | 13 | 2135 | 8861224 |