Gene Transcripts

Gene Symbol
Hmgb4
Entrez Gene ID
69317
Gene Name
high-mobility group box 4
Gene Aliases
1700001F22Rik, AI326135
Chromosome Location
Chr. 4: 128260212 - 128260895 on Build GRCm38
UniGene ID
Mm.331893
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027036.3 NP_081312.2 1 512
GenBank mRNA AK005588.1 - 1 511
BC048744.1 - 2 472
BC061030.1 - 1 508 6775221
BC087910.1 - 2 398
BC087934.1 - 2 468
CA465179.1 - 1 511

View Full Product Details