Gene Transcripts

Gene Symbol
Ttc9
Entrez Gene ID
69480
Gene Name
tetratricopeptide repeat domain 9
Gene Aliases
1700029M07Rik, AI429215, AI853559
Chromosome Location
Chr. 12: 81629771 - 81664941 on Build GRCm38
UniGene ID
Mm.130002
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033149.2 NP_001028321.1 2 579
GenBank mRNA AK133456.1 - 2 580
AK161499.1 - 2 379
BC139242.1 - 2 577 8862264
BC139244.1 - 2 577 8862266

View Full Product Details