Gene Transcripts

Gene Symbol
CD200
Entrez Gene ID
4345
Gene Name
CD200 molecule
Gene Aliases
MOX1, MOX2, MRC, OX-2
Chromosome Location
Chr. 3: 112332224 - 112362812 on Build GRCh38
UniGene ID
Hs.79015
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004196.3 NP_001004196.2 4, 5 756
NM_001318828.1 NP_001305757.1 2, 3 599
NM_005944.6 NP_005935.4 3, 4 681
XM_005247481.4 XP_005247538.1 4, 5 930
XM_005247482.2 XP_005247539.1 3, 4 560
XM_005247485.2 XP_005247542.1 3, 4 871
XM_005247486.2 XP_005247543.1 2, 3 368
XM_017006452.1 XP_016861941.1 4, 5 1257
XM_017006453.1 XP_016861942.1 2, 3 523
XM_017006454.1 XP_016861943.1 4, 5 1246
XM_017006455.1 XP_016861944.1 3, 4 881
XM_017006456.1 XP_016861945.1 4, 5 607
GenBank mRNA AF063591.1 - 3, 4 472
AK074972.1 - 3, 4 446
AK293399.1 - 2, 3 368
AK297194.1 - 4, 5 547
AL832529.1 - 3, 4 466
AY603771.1 - 2, 3 415
BC022522.1 - 3, 4 474 4794050
BC031103.1 - 3, 4 450 5301702
BI547787.1 - 4, 5 552

View Full Product Details