Gene Transcripts

Gene Symbol
Cyp2u1
Entrez Gene ID
71519
Gene Name
cytochrome P450, family 2, subfamily u, polypeptide 1
Gene Aliases
8430436A10Rik
Chromosome Location
Chr. 3: 131290491 - 131303227 on Build GRCm38
UniGene ID
Mm.473179
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027816.3 NP_082092.2 2 797
GenBank mRNA AK018458.1 - 2 773
AK041477.1 - 2 798
AK142740.1 - 2 737

View Full Product Details