Gene Transcripts

Gene Symbol
Dhdh
Entrez Gene ID
71755
Gene Name
dihydrodiol dehydrogenase (dimeric)
Gene Aliases
1300018L09Rik, AA591799, Xld-1, Xld1
Chromosome Location
Chr. 7: 45473563 - 45488873 on Build GRCm38
UniGene ID
Mm.34208
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027903.3 NP_082179.1 4 498
GenBank mRNA AK005050.2 - 4 499
AK154452.1 - 4 480
BC031710.1 - 4 476
BC116414.2 - 4 482 40103176

View Full Product Details