Gene Transcripts

Gene Symbol
Nupl1
Entrez Gene ID
71844
Gene Name
nucleoporin like 1
Gene Aliases
1700017F11Rik, mKIAA0410
Chromosome Location
Chr. 14: 60205610 - 60251502 on Build GRCm38
UniGene ID
Mm.258051
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_170591.1 NP_733479.1 4 526
GenBank mRNA AK006062.1 - 4 628
AK029688.1 - 580
AK078279.1 - 4 574
AK086412.1 - 4 594
AK133721.1 - 4 537
AK139895.1 - 4 657
AK146477.1 - 4 526
AK163792.1 - 4 555
BC026743.1 - 4 557 3964978

View Full Product Details