Gene Transcripts

Gene Symbol
Fbxl20
Entrez Gene ID
72194
Gene Name
F-box and leucine-rich repeat protein 20
Gene Aliases
2610511F20Rik, 4632423N09Rik, AI849362, AL117906, C86145, Fbl2, Scr, mKIAA4147
Chromosome Location
Chr. 11: 98082554 - 98150440 on Build GRCm38
UniGene ID
Mm.218350
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028149.1 NP_082425.1 7 722
GenBank mRNA AK012109.1 - 7 446
AK144786.1 - 7 696
AK220232.1 - 7 675
EF649694.1 - 7 469

View Full Product Details