Gene Transcripts

Gene Symbol
Rnf38
Entrez Gene ID
73469
Gene Name
ring finger protein 38
Gene Aliases
1700065B19Rik, 2610202O07Rik, AA673263, Oip1
Chromosome Location
Chr. 4: 44126212 - 44233643 on Build GRCm38
UniGene ID
Mm.262859
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001038993.3 NP_001034082.1 8 1142
NM_175201.5 NP_780410.2 7 1172
GenBank mRNA AK006888.1 - 7 1133
AK147608.1 - 8 1199
AK163346.1 - 3 303
BC060730.1 - 7 1149 6408623
BC062976.1 - 7 1086 6848803

View Full Product Details