Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_028773.3 | NP_083049.1 | 2 | 259 | |
GenBank mRNA | AJ306422.1 | - | 2 | 125 | |
AK004734.2 | - | 2 | 260 | ||
AK156202.1 | - | 2 | 324 | ||
AK171017.1 | - | 2 | 257 | ||
BC064734.1 | - | 2 | 244 |