Gene Transcripts

Gene Symbol
NAP1L4
Entrez Gene ID
4676
Gene Name
nucleosome assembly protein 1 like 4
Gene Aliases
NAP1L4b, NAP2, NAP2L, hNAP2
Chromosome Location
Chr. 11: 2944430 - 2992377 on Build GRCh38
UniGene ID
Hs.731784
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005969.3 NP_005960.1 9, 10 895
GenBank mRNA AB500094.1 - 7, 8 754
AK299515.1 - 9, 10 818
AK312599.1 - 9, 10 903
AK316242.1 - 9, 10 812
AK316324.1 - 9, 10 833
AK316548.1 - 10, 11 952
BC022090.2 - 9, 10 805 3029455
U77456.1 - 9, 10 903

View Full Product Details