Gene Transcripts
- Gene Symbol
- Sf3a3
- Entrez Gene ID
- 75062
- Gene Name
- splicing factor 3a, subunit 3
- Gene Aliases
- 4930512K19Rik, 60kDa
- Chromosome Location
- Chr. 4: 124714861 - 124732423 on Build GRCm38
- UniGene ID
- Mm.25779
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_029157.3 |
NP_083433.1
|
11, 12 |
968 |
|
GenBank mRNA |
AK030743.1 |
- |
11, 12 |
1003 |
|
AK150733.1 |
- |
11, 12 |
966 |
|
AK151141.1 |
- |
11, 12 |
966 |
|
AK151382.1 |
- |
11, 12 |
966 |
|
AK161646.1 |
- |
11, 12 |
989 |
|
AK168966.1 |
- |
11, 12 |
992 |
|
AK169324.1 |
- |
11, 12 |
1012 |
|
AK187073.1 |
- |
4, 5 |
368 |
|
AK213498.1 |
- |
1, 2 |
110 |
|
AK215652.1 |
- |
1, 2 |
110 |
|
BC009141.1 |
- |
11, 12 |
968 |
3598647 |
BC092058.1 |
- |
11, 12 |
1000 |
4017388 |
View Full Product Details