Gene Transcripts

Gene Symbol
Slitrk5
Entrez Gene ID
75409
Gene Name
SLIT and NTRK-like family, member 5
Gene Aliases
2610019D03Rik
Chromosome Location
Chr. 14: 111675115 - 111683144 on Build GRCm38
UniGene ID
Mm.235097
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198865.1 NP_942565.1 2 1204
GenBank mRNA AK173062.1 - 1 235
BC065107.1 - 2 823 30357422

View Full Product Details