Gene Transcripts

Gene Symbol
NDUFB9
Entrez Gene ID
4715
Gene Name
NADH:ubiquinone oxidoreductase subunit B9
Gene Aliases
B22, CI-B22, LYRM3, UQOR22
Chromosome Location
Chr. 8: 124539102 - 124549986 on Build GRCh38
UniGene ID
Hs.15977
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278645.1 NP_001265574.1 4 501
NM_001278646.1 NP_001265575.1 4 534
NM_001311168.1 NP_001298097.1 4 503
NM_005005.2 NP_004996.1 4 536
GenBank mRNA AF044956.1 - 4 530
AF067168.1 - 4 530
AF261090.1 - 4 535
AI907018.1 - 4 553
AK026538.1 - 4 519
AK313432.1 - 4 498
AL136714.1 - 4 506
BC007672.2 - 4 503 3160562
BG703246.1 - 4 463
BQ685604.1 - 4 486
CN410734.1 - 4 444
DB504123.1 - 4 461
S82655.1 - 3 222

View Full Product Details