Gene Transcripts

Gene Symbol
Rilpl1
Entrez Gene ID
75695
Gene Name
Rab interacting lysosomal protein-like 1
Gene Aliases
2900002H16Rik, 6330559I19Rik, GOSPEL
Chromosome Location
Chr. 5: 124493080 - 124531391 on Build GRCm38
UniGene ID
Mm.41180
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021430.2 NP_067405.1 6 1409
GenBank mRNA AB041584.1 - 6 1039
AK013469.1 - 3 302
BC029965.1 - 6 1087
BC051945.1 - 6 1409 5716540
BC051946.1 - 6 1339 6403995
BC058384.1 - 6 1302 5718399

View Full Product Details