Gene Transcripts

Gene Symbol
Osbp
Entrez Gene ID
76303
Gene Name
oxysterol binding protein
Gene Aliases
1110018F06Rik, AW559088, mKIAA4220
Chromosome Location
Chr. 19: 11965844 - 11994114 on Build GRCm38
UniGene ID
Mm.291279
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033174.1 NP_001028346.1 4 984
GenBank mRNA AK132663.1 - 2 264
AK220301.1 - 1 22
BC107327.1 - 4 718
BC107328.2 - 4 718

View Full Product Details