Gene Transcripts

Gene Symbol
Cluap1
Entrez Gene ID
76779
Gene Name
clusterin associated protein 1
Gene Aliases
2310030D15Rik, 2610111M03Rik
Chromosome Location
Chr. 16: 3907659 - 3941147 on Build GRCm38
UniGene ID
Mm.458921
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029738.2 NP_084014.1 12 1324
GenBank mRNA AK134648.1 - 12 1291
BC024910.1 - 12 1324 3603454
BC063769.1 - 6 558

View Full Product Details