Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_199065.2 | NP_951020.1 | 1 | 2117 | |
GenBank mRNA | AK014285.1 | - | 1 | 53 | |
AK144290.1 | - | 1 | 1968 | ||
AK173296.1 | - | 2036 | |||
BC150756.1 | - | 1 | 1225 | 9087667 | |
BC157902.1 | - | 1 | 1225 | 9007276 |