Gene Transcripts

Gene Symbol
Lrrc42
Entrez Gene ID
77809
Gene Name
leucine rich repeat containing 42
Gene Aliases
A930011F22Rik, AA536996
Chromosome Location
Chr. 4: 107233514 - 107253533 on Build GRCm38
UniGene ID
Mm.29681
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029985.2 NP_084261.1 2 323
GenBank mRNA AK020847.1 - 2 129
AK143870.1 - 1 97
BC027203.1 - 2 255 3710091
BC031894.1 - 2 227 5362699
CJ174174.1 - 2 324

View Full Product Details