Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006166.3 | NP_006157.1 | 5 | 544 | |
XM_005268908.4 | XP_005268965.1 | 6 | 608 | ||
XM_005268909.4 | XP_005268966.1 | 6 | 591 | ||
XM_005268910.4 | XP_005268967.1 | 6 | 1263 | ||
GenBank mRNA | AK291934.1 | - | 5 | 476 | |
BC005316.1 | - | 5 | 493 | 3680937 | |
BC005317.1 | - | 5 | 499 | 3997299 | |
BC007035.1 | - | 5 | 471 | 3996244 | |
BG776259.1 | - | 5 | 545 | ||
L06145.1 | - | 3 | 343 | ||
X59710.1 | - | 5 | 448 |