Gene Transcripts

Gene Symbol
Troap
Entrez Gene ID
78733
Gene Name
trophinin associated protein
Gene Aliases
AW476063, E130301L11Rik, tastin
Chromosome Location
Chr. 15: 99074973 - 99083409 on Build GRCm38
UniGene ID
Mm.124905
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001162506.1 NP_001155978.1 3 310
NM_030159.1 NP_084435.1 3 335
GenBank mRNA AK021408.1 - 3 322
AK050381.1 - 3 331
AK077509.1 - 3 300
BC080746.1 - 3 295
BC100405.1 - 3 264
BC145227.1 - 3 708 9053779

View Full Product Details