Gene Transcripts

Gene Symbol
Gins3
Entrez Gene ID
78833
Gene Name
GINS complex subunit 3 (Psf3 homolog)
Gene Aliases
2700085M18Rik, AI616142
Chromosome Location
Chr. 8: 95633559 - 95645059 on Build GRCm38
UniGene ID
Mm.35546
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030198.3 NP_084474.1 2, 3 466
GenBank mRNA AK019225.1 - 2, 3 467
AK146574.1 - 2, 3 497
AK150602.1 - 2, 3 498
AK151243.1 - 2, 3 498
AK152572.1 - 2, 3 498
AK160606.1 - 2, 3 467
BC039794.1 - 2, 3 463 5121680
BC064746.1 - 2, 3 455 30095310

View Full Product Details