Gene Transcripts

Gene Symbol
Pik3ap1
Entrez Gene ID
83490
Gene Name
phosphoinositide-3-kinase adaptor protein 1
Gene Aliases
1810044J04Rik, BCAP
Chromosome Location
Chr. 19: 41272752 - 41385070 on Build GRCm38
UniGene ID
Mm.222266
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031376.3 NP_113553.1 2 478
GenBank mRNA AF293806.1 - 2 478
AK087295.1 - 2 505
AK089785.1 - 2 511
AK154326.1 - 2 506
AK170130.1 - 2 511
AK170925.1 - 2 511
AK171021.1 - 2 506
BC113141.1 - 2 437 40047744

View Full Product Details