Gene Transcripts

Gene Symbol
Mrpl43
Entrez Gene ID
94067
Gene Name
mitochondrial ribosomal protein L43
Gene Aliases
4930442D21Rik, bMRP36a
Chromosome Location
Chr. 19: 45005014 - 45006442 on Build GRCm38
UniGene ID
Mm.44174
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053164.3 NP_444394.1 3 382
GenBank mRNA AB049657.1 - 3 343
AK015357.1 - 3 363
AK076204.1 - 3 341
BC027676.1 - 3 322
BC048733.1 - 3 373 6512868
BC065812.1 - 3 329
BY251956.1 - 3 383

View Full Product Details