Gene Transcripts
- Gene Symbol
- Osbpl11
- Entrez Gene ID
- 106326
- Gene Name
- oxysterol binding protein-like 11
- Gene Aliases
- 9430097N02Rik, AI132306, AU019795, ORP-11
- Chromosome Location
- Chr. 16: 33169763 - 33243312 on Build GRCm38
- UniGene ID
- Mm.486405
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_176840.3 |
NP_789810.2
|
10 |
2570 |
|
GenBank mRNA |
BC035278.1 |
- |
10 |
2570 |
|
View Full Product Details