Gene Transcripts

Gene Symbol
OR1F1
Entrez Gene ID
4992
Gene Name
olfactory receptor family 1 subfamily F member 1
Gene Aliases
OLFMF, OR16-36, OR16-37, OR16-88, OR16-89, OR16-90, OR1F10, OR1F13P, OR1F4, OR1F5, OR1F6, OR1F7, OR1F8, OR1F9, OR3-145, ORL1023
Chromosome Location
Chr. 16: 3188195 - 3206559 on Build GRCh38
UniGene ID
Hs.664502
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012360.1 NP_036492.1 1 595
XM_011522506.2 XP_011520808.1 4 1042
XM_011522507.2 XP_011520809.1 2 1465
GenBank mRNA BC069335.1 - 1 595
Y14442.1 - 1 595

View Full Product Details