Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_181404.5 | NP_852069.4 | 6, 7 | 3328 | |
GenBank mRNA | AK122214.1 | - | 2, 3 | 1694 | |
AK147945.1 | - | 3, 4 | 2915 | ||
BC050250.1 | - | 1, 2 | 134 | ||
BC058673.1 | - | 3, 4 | 1112 | ||
BC079563.1 | - | 2, 3 | 2655 | 30535512 |