Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027664.1 | NP_081940.1 | 3 | 525 | |
GenBank mRNA | AK016936.1 | - | 2 | 251 | |
AK039398.1 | - | 2 | 199 | ||
AK173135.1 | - | 4 | 535 | ||
BB616179.1 | - | 3 | 726 | ||
BC052078.1 | - | 3 | 497 | 6402765 |