Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027664.1 | NP_081940.1 | 6 | 1058 | |
GenBank mRNA | AK016936.1 | - | 5 | 784 | |
AK039398.1 | - | 5 | 732 | ||
AK173135.1 | - | 7 | 1068 | ||
BC052078.1 | - | 6 | 1030 | 6402765 |