Gene Transcripts
- Gene Symbol
- Pttg1ip
- Entrez Gene ID
- 108705
- Gene Name
- pituitary tumor-transforming 1 interacting protein
- Gene Aliases
- 1810010L20Rik, AI314311, AU018448, C79540
- Chromosome Location
- Chr. 10: 77581767 - 77598732 on Build GRCm38
- UniGene ID
- Mm.28853
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_145925.2 |
NP_666037.1
|
1, 2 |
161 |
|
GenBank mRNA |
AK035696.1 |
- |
1, 2 |
176 |
|
AK075773.1 |
- |
1, 2 |
134 |
|
AK088813.1 |
- |
1, 2 |
198 |
|
AK089887.1 |
- |
1, 2 |
143 |
|
AK098093.1 |
- |
1, 2 |
175 |
|
AK151639.1 |
- |
1, 2 |
143 |
|
AK152366.1 |
- |
1, 2 |
143 |
|
AK159987.1 |
- |
1, 2 |
143 |
|
AK167972.1 |
- |
1, 2 |
173 |
|
AK168164.1 |
- |
1, 2 |
173 |
|
AK170802.1 |
- |
1, 2 |
143 |
|
AK170971.1 |
- |
1, 2 |
176 |
|
AK170972.1 |
- |
1, 2 |
143 |
|
AK171140.1 |
- |
1, 2 |
142 |
|
BC023961.1 |
- |
1, 2 |
216 |
5323397 |
BC025533.1 |
- |
1, 2 |
185 |
5250502 |
BC029144.1 |
- |
1, 2 |
157 |
4945974 |
BC032184.1 |
- |
1, 2 |
180 |
5310191 |
View Full Product Details