Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_175382.3 | NP_780591.1 | 2 | 930 | |
GenBank mRNA | AK041641.1 | - | 2 | 932 | |
AK076058.1 | - | 2 | 952 | ||
BC090647.1 | - | 2 | 1016 | 30633261 | |
BC106123.1 | - | 2 | 914 | 3995296 | |
BC138225.1 | - | 2 | 899 | 8861244 | |
BC145667.1 | - | 2 | 899 | 40130970 |