Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_033612.2 | NP_291090.2 | 4, 5 | 370 | |
GenBank mRNA | AK007392.1 | - | 4, 5 | 368 | |
BC011218.1 | - | 4, 5 | 344 | 4208442 | |
CB951068.1 | - | 4, 5 | 370 | ||
M27347.1 | - | 3, 4 | 253 |