Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000430.3 | NP_000421.1 | 3 | 623 | |
XM_011523901.1 | XP_011522203.1 | 4 | 672 | ||
XM_011523902.2 | XP_011522204.1 | 4 | 504 | ||
XM_011523903.2 | XP_011522205.1 | 4 | 422 | ||
XM_017024699.1 | XP_016880188.1 | 3 | 373 | ||
XM_017024700.1 | XP_016880189.1 | 3 | 450 | ||
XM_017024701.1 | XP_016880190.1 | 3 | 366 | ||
GenBank mRNA | AF208835.1 | - | 2 | 55 | |
AF208836.1 | - | 2 | 55 | ||
AF400434.1 | - | 2 | 248 | ||
AK313078.1 | - | 3 | 277 | ||
BC064638.1 | - | 3 | 265 | 5786560 | |
L13385.1 | - | 3 | 272 | ||
L13386.1 | - | 3 | 508 |