Gene Transcripts
- Gene Symbol
- C79798
- Entrez Gene ID
- 96990
- Gene Name
- expressed sequence C79798
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.441760
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK138624.1 |
- |
2 |
210 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB633614.1 |
- |
2 |
210 |
|
Chr. 2 - 34287771 - 34303082 [+] |
GenBankmRNA |
BY724378.1 |
- |
2 |
210 |
|
Chr. 2 - 34287771 - 34303264 [+] |
View Full Product Details