Gene Transcripts
- Gene Symbol
- C79798
- Entrez Gene ID
- 96990
- Gene Name
- expressed sequence C79798
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.441760
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK138624.1 |
- |
2 |
1550 |
|
BG079793.2 |
- |
1 |
569 |
|
View Full Product Details