Gene Transcripts
- Gene Symbol
- CAV3
- Entrez Gene ID
- 859
- Gene Name
- caveolin 3
- Gene Aliases
- LGMD1C, LQT9, VIP-21, VIP21
- Chromosome Location
- Chr. 3: 8733800 - 8746765 on Build GRCh38
- UniGene ID
- Hs.98303
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001234.4 |
NP_001225.1
|
1, 2 |
190 |
|
NM_033337.2 |
NP_203123.1
|
1, 2 |
190 |
|
GenBank mRNA |
AF036365.1 |
- |
1, 2 |
160 |
|
AF043101.1 |
- |
1, 2 |
110 |
|
AK291892.1 |
- |
1, 2 |
190 |
|
BC069368.1 |
- |
1, 2 |
163 |
7262168 |
BC102033.1 |
- |
1, 2 |
150 |
40033505 |
BC102036.1 |
- |
1, 2 |
150 |
40033114 |
BC102037.1 |
- |
1, 2 |
150 |
40033118 |
Y14747.1 |
- |
1, 2 |
124 |
|
View Full Product Details