Gene Transcripts
- Gene Symbol
- Cyb5b
- Entrez Gene ID
- 66427
- Gene Name
- cytochrome b5 type B
- Gene Aliases
- 1810044O22Rik, AU015618, Cyb5m
- Chromosome Location
- Chr. 8: 107150661 - 107187470 on Build GRCm38
- UniGene ID
- Mm.20242
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_025558.5 |
NP_079834.2
|
3, 4 |
369 |
|
GenBank mRNA |
AK003333.2 |
- |
3, 4 |
370 |
|
AK003815.1 |
- |
3, 4 |
359 |
|
AK007780.1 |
- |
|
389 |
|
AK019263.1 |
- |
3, 4 |
369 |
|
AK052810.1 |
- |
3, 4 |
370 |
|
AK083359.1 |
- |
3, 4 |
215 |
|
AK088970.1 |
- |
3, 4 |
371 |
|
AK165543.1 |
- |
3, 4 |
373 |
|
AK166544.1 |
- |
3, 4 |
347 |
|
AK168422.1 |
- |
3, 4 |
359 |
|
BC046479.1 |
- |
3, 4 |
332 |
|
BC046810.1 |
- |
3, 4 |
335 |
|
BC048147.1 |
- |
3, 4 |
359 |
|
BC054749.1 |
- |
3, 4 |
390 |
6410090 |
BC058812.1 |
- |
3, 4 |
332 |
6530835 |
BC062980.1 |
- |
3, 4 |
381 |
6842939 |
View Full Product Details