Gene Transcripts

Gene Symbol
Cabp4
Entrez Gene ID
73660
Gene Name
calcium binding protein 4
Gene Aliases
2410038D05Rik
Chromosome Location
Chr. 19: 4135423 - 4144080 on Build GRCm38
UniGene ID
Mm.379226
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144532.2 NP_653115.1 1, 2 400
GenBank mRNA AK171863.1 - 5, 6 1010
AY039218.1 - 1, 2 395
BC029002.1 - 1, 2 308
BC049263.1 - 1, 2 400 5366999
BC053330.1 - 1, 2 355

View Full Product Details