Gene Transcripts

Gene Symbol
Dcun1d4
Entrez Gene ID
100737
Gene Name
DCN1, defective in cullin neddylation 1, domain containing 4 (S. cerevisiae)
Gene Aliases
AI836376
Chromosome Location
Chr. 5: 73481027 - 73560794 on Build GRCm38
UniGene ID
Mm.220312
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001190733.1 NP_001177662.1 4 340
NM_001190734.1 NP_001177663.1 5 275
NM_178896.5 NP_849227.1 4 340
GenBank mRNA AK033559.1 - 4 285
AK076991.1 - 5 276
BC060213.1 - 4 281
BC108346.1 - 4 242 6310392
BC137627.1 - 4 284 8860646
BC137628.1 - 4 284 8860647
BC144794.1 - 4 284 9053342

View Full Product Details