Gene Transcripts
- Gene Symbol
- Klra6
- Entrez Gene ID
- 16637
- Gene Name
- killer cell lectin-like receptor, subfamily A, member 6
- Gene Aliases
- Ly49f, ly-49f
- Chromosome Location
- Chr. 6: 130013033 - 130026960 on Build GRCm38
- UniGene ID
- Mm.14112
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_008464.2 |
NP_032490.1
|
2, 3 |
202 |
|
GenBank mRNA |
U10092.1 |
- |
2, 3 |
202 |
|
View Full Product Details