Gene Transcripts
- Gene Symbol
- AKR1B1
- Entrez Gene ID
- 231
- Gene Name
- aldo-keto reductase family 1 member B
- Gene Aliases
- ADR, ALDR1, ALR2, AR
- Chromosome Location
- Chr. 7: 134442352 - 134459136 on Build GRCh38
- UniGene ID
- Hs.521212
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001628.2 |
NP_001619.1
|
8, 9 |
907 |
|
XM_005250234.3 |
XP_005250291.1
|
8, 9 |
916 |
|
GenBank mRNA |
AB209796.1 |
- |
7, 8 |
1424 |
|
AF328729.1 |
- |
8, 9 |
848 |
|
AK313439.1 |
- |
8, 9 |
898 |
|
BC000260.1 |
- |
8, 9 |
907 |
3357652 |
BC005387.1 |
- |
8, 9 |
871 |
3960811 |
BC010391.1 |
- |
8, 9 |
873 |
4049736 |
BT019859.1 |
- |
8, 9 |
833 |
|
CR450351.1 |
- |
8, 9 |
833 |
|
CR542203.1 |
- |
8, 9 |
833 |
|
J04795.1 |
- |
8, 9 |
842 |
|
J05017.1 |
- |
8, 9 |
869 |
|
J05474.1 |
- |
8, 9 |
865 |
|
M34720.1 |
- |
8, 9 |
846 |
|
X15414.1 |
- |
8, 9 |
878 |
|
View Full Product Details